missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Chromium wire, Alfa Aesar™

Form: approx. 0.43g/m

Manufacturer:  Thermo Fisher Scientific 44187H9

 View more versions of this product

Catalog No. AA44187H9



Specifications

Specifications

Nickel Chromium
11106-97-1
CrNi
acmc-1byoe
[Cr].[Ni]
Odorless
11320920
Silver
500 m
Silver-colored wire
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
chromium;nickel
110.689
Wire, ≈0.017g/m

Chemical Identifiers

11106-97-1
110.689
VNNRSPGTAMTISX-UHFFFAOYSA-N
11320920
[Cr].[Ni]
CrNi
MFCD00673110
acmc-1byoe
chromium;nickel
Safety and Handling

Safety and Handling

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents