Learn More
Nickel Chromium wire, 0.51mm (0.02in) dia, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 013967H4
Description
These alloys are typically used to increase the resistivity of wire. They are also used in some dental restorations (fillings). It is commonly used in ceramic as an internal support. Nichrome wire is used for its ability to withstand the high temperatures that occur when clay work is fired in a kiln.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Chromium wire | |
11106-97-1 | |
CrNi | |
100 m | |
Insoluble in water. | |
[Cr].[Ni] | |
110.69 | |
Odorless | |
Wire |
0.51mm (0.02 in.) dia. | |
Ni:Cr; 80:20 wt% | |
MFCD00673110 | |
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
VNNRSPGTAMTISX-UHFFFAOYSA-N | |
chromium;nickel | |
11320920 | |
≈1.86g/m |
Chemical Identifiers
11106-97-1 | |
110.69 | |
VNNRSPGTAMTISX-UHFFFAOYSA-N | |
11320920 |
CrNi | |
MFCD00673110 | |
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
[Cr].[Ni] |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only