missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Chromium wire, 0.143mm (0.0056 in.) dia.

High resistivity material for electronics industry

Manufacturer:  Alfa Aesar™ 13966J5

 View more versions of this product

Catalog No. AA13966J5



Specifications

Specifications

Ni:Cr; 80:20 wt%
≈0.14g/m
0.143mm (0.0056 in.) dia.
CrNi
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
[Cr].[Ni]
Odorless
11320920
Nickel Chromium wire
2000m
11106-97-1
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
chromium;nickel
110.689
Wire

Chemical Identifiers

11106-97-1
110.689
VNNRSPGTAMTISX-UHFFFAOYSA-N
11320920
[Cr].[Ni]
CrNi
MFCD00673110
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
chromium;nickel
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents