Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Chromium, sputtering target, 76.2in (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

$349.57 - $349.57

Chemical Identifiers

Chemical Name or Material Nickel Chromium, sputtering target
CAS 11106-97-1
Name Note 76.2in (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00673110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40872KS

  SDS
alfa aesar™
40872KS
1 each Each for $349.57
Description & Specifications

Specifications

Odor Odorless
Health Hazard 2 GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00673110
Health Hazard 1 Warning
Assay 99.95%
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Chemical Name or Material Nickel Chromium, sputtering target
TSCA TSCA
Quantity 1 each
Assay Percent Notes (metals basis)
Name Note 76.2in (3.0 in.) dia. x 3.18mm (0.125 in.) thick
CAS 11106-97-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.