missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$266.27 - $266.27

Chemical Identifiers

CAS 11106-97-1
MDL Number MFCD00673110
Chemical Name or Material Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40870KS SDS Alfa Aesar™
40870KS
1 each Each for $266.27
Specifications Description & Specifications

Chemical Identifiers

11106-97-1
MFCD00673110
Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

Specifications

Ni:Cr; 80:20 wt%
11106-97-1
Odorless
99.95%, (metals basis)
1 each
MFCD00673110
Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA