Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Chromium powder, -140+325 mesh, 97% (metals basis)

Chemical Identifiers

Chemical Name or Material Nickel Chromium powder
CAS 11106-97-1
MDL Number MFCD00673110
UN Number UN3178
Synonym Nichrome™
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1310836

  SDS
alfa aesar™
1310836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nickel Chromium powder
TSCA TSCA
Density 3.1g/mL
Physical Form Powder
Assay 97%
Assay Percent Notes (metals basis)
MDL Number MFCD00673110
CAS 11106-97-1
UN Number UN3178
Mesh Size −140+325 Mesh
Synonym Nichrome™
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 500g