Please login to your online account to display your discounted pricing

Nickel Chromium gauze, Alfa Aesar™

Open area: 36%; Width of opening: 0.015 in.

Chemical Identifiers

Chemical Name or Material Nickel Chromium
Name Note gauze
CAS 11106-97-1
Physical Form Wire Cloth
Molecular Formula Ni:Cr; 80:20 wt%
MDL Number MFCD00673110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40941FL

 
alfa aesar
40941FL
75 x 75mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40941GT

 
alfa aesar
40941GT
150 x 150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40941HB

 
alfa aesar
40941HB
300 x 300mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Wire Cloth
Quantity 75 x 75mm
Color Silver
Chemical Name or Material Nickel Chromium
Odor Odorless
Name Note gauze
Molecular Formula Ni:Cr; 80:20 wt%
MDL Number MFCD00673110
Appearance Silver -colored gauze
CAS 11106-97-1
Health Hazard 1 H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.
Safety and Handling

H351-H317: Suspected of causing cancer. May cause an allergic skin reaction.