missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Chromium Aluminum Copper wire, 0.025mm (0.001 in.) dia., annealed (Evanohm™)

Specifications

Chemical Name or Material Nickel Chromium Aluminum Copper wire
Name Note 0.025mm (0.001 in.) dia., Annealed (Evanohm™)
MDL Number MFCD02684522
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43635G9 View Documents Alfa Aesar™
43635G9
25m N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA43635G5 View Documents Alfa Aesar™
43635G5
5m N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Nickel Chromium Aluminum Copper wire
0.025mm (0.001 in.) dia., Annealed (Evanohm™)
MFCD02684522