Learn More
Nickel chromite, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01942414
Description
In mixing Nickel (II) Chromate with Aluminates or Cobalt Chromate, a binary compound known as a spinel is formed that is able to withstand temperatures up to 1000Â °C. The products from these mixings can be used as oxidation-resistive components in airplane and space shuttle coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel chromite | |
Powder | |
NiCr2O4 | |
Insoluble in water. Soluble in hydrochloric acid. | |
Odorless |
12018-18-7 | |
25 g | |
MFCD00064806 | |
226.7 |
Chemical Identifiers
12018-18-7 | |
MFCD00064806 |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
EINECSNumber : 234-636-7
RTECSNumber : GB9000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only