Alfa Aesar™ Nickel chromite
Manufacturer:  Alfa Aesar™ 1942422
 View more versions of this product
Catalog No. AA1942422
Specifications
Specifications
Nickel chromite
NiCr2O4
226.7
−100 Mesh
100g
12018-18-7
MFCD00064806
Powder
Odorless
Chemical Identifiers
12018-18-7
MFCD00064806
Safety and Handling
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
RTECSNumber : GB9000000
TSCA : TSCA
SDS Documents