Please login to your online account to display your discounted pricing

Nickel Carbonate (Powder/Technical), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  N51500

 View more versions of this product

Catalog No. N51-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 118.7
Quantity 500g
Physical Form Solid
Viscosity [NULL]
Color Light Green
Density 2.6g/cm³
Chemical Name or Material Nickel Carbonate
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00011144
CAS 3333-67-3
ChemAlert Storage Symbol Blue
Synonym Nickel (II) Carbonate
Molecular Formula NiCO3

Specifications

Chemical Name or Material Nickel Carbonate
CAS 3333-67-3
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula NiCO3
Formula Weight 118.7
MDL Number MFCD00011144
Synonym Nickel (II) Carbonate
Accessories
 

SDS & Safety Documents