Please login to your online account to display your discounted pricing

Nickel Carbonate (Powder/Technical), Fisher Chemical

$180.20 - $610.40

Chemical Identifiers

Chemical Name or Material Nickel Carbonate
CAS 3333-67-3
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula NiCO3
Formula Weight 118.7
MDL Number MFCD00011144
Synonym Nickel (II) Carbonate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

N51-500

  SDS
fisher scientific company
N51500
Poly Bottle 500g Each for $610.40
Case of 6 Each for $3,662.40

N51-100

  SDS
fisher scientific company
N51100
Glass Bottle 100g Each for $180.20
Case of 6 Each for $1,081.20
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 118.7
Quantity 500g
Physical Form Solid
Color Light Green
Density 2.6g/cm³
Chemical Name or Material Nickel Carbonate
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00011144
CAS 3333-67-3
ChemAlert Storage Symbol Blue
Synonym Nickel (II) Carbonate
Molecular Formula NiCO3
Safety and Handling