Learn More
Nickel boride, 99% (metals basis), Thermo Scientific Chemicals
$514.78 - $514.78
Chemical Identifiers
CAS | 12007-00-0 |
---|---|
Molecular Formula | BNi |
Molecular Weight (g/mol) | 69.503 |
MDL Number | MFCD00016261 |
InChI Key | QDWJUBJKEHXSMT-UHFFFAOYSA-N |
Synonym | nickel boride nib, nickelylidyneborane, nickel boride, powder,-35 mesh trace metals basis |
PubChem CID | 82791 |
IUPAC Name | boranylidynenickel |
SMILES | B#[Ni] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1311022
|
Thermo Scientific Chemicals
01311022 |
100 g |
Each for $514.78
|
|
|||||
Description
Nickel boride is an efficient catalyst and reducing agent. It is used as a heterogeneous hydrogenation catalyst. It can also be used to cleave thioacetals. It is used in a protection-deprotection scheme for triterpenoid ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12007-00-0 | |
69.503 | |
QDWJUBJKEHXSMT-UHFFFAOYSA-N | |
82791 | |
B#[Ni] |
BNi | |
MFCD00016261 | |
nickel boride nib, nickelylidyneborane, nickel boride, powder,-35 mesh trace metals basis | |
boranylidynenickel |
Specifications
12007-00-0 | |
Powder | |
BNi | |
UN3077 | |
Insoluble in water. | |
B#[Ni] | |
69.503 | |
69.52 | |
Odorless | |
Nickel boride |
White to Gray | |
100 g | |
MFCD00016261 | |
nickel boride nib, nickelylidyneborane, nickel boride, powder,-35 mesh trace metals basis | |
QDWJUBJKEHXSMT-UHFFFAOYSA-N | |
boranylidynenickel | |
82791 | |
99% | |
7.39 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 234-493-0
TSCA : Yes
RUO – Research Use Only