missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel boride, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1311022

 View more versions of this product

Catalog No. AA1311022



Specifications

Specifications

Nickel boride
(metals basis)
MFCD00016261
nickel boride nib, nickelylidyneborane, nickel boride, powder,-35 mesh trace metals basis
B#[Ni]
69.503
69.52
99%
−325 Mesh
White to Gray
12007-00-0
BNi
UN3077
QDWJUBJKEHXSMT-UHFFFAOYSA-N
boranylidynenickel
82791
Powder
7.39g/mL
Odorless
100g

Chemical Identifiers

12007-00-0
69.503
QDWJUBJKEHXSMT-UHFFFAOYSA-N
82791
B#[Ni]
BNi
MFCD00016261
nickel boride nib, nickelylidyneborane, nickel boride, powder,-35 mesh trace metals basis
boranylidynenickel
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Documents