Please login to your online account to display your discounted pricing

Nickel boride, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nickel boride
CAS 12007-01-1
Physical Form -35 Mesh Granular
Assay Percent Range 99%
Molecular Formula Ni2B
Formula Weight 128.23
MDL Number MFCD00084757
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1310914

 
alfa aesar
1310914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1310922

 
alfa aesar
1310922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.23
Quantity 25g
Chemical Name or Material Nickel boride
EINECS Number 234-494-6
Physical Form -35 Mesh Granular
Assay Percent Range 99%
Molecular Formula Ni2B
MDL Number MFCD00084757
CAS 12007-01-1