Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel arsenide, 99.5% (metals basis excluding Co), Co 0.1-1%

Chemical Identifiers

Chemical Name or Material Nickel arsenide
CAS 12068-61-0
MDL Number MFCD00151450
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3626906

  SDS
alfa aesar™
3626906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3626914

  SDS
alfa aesar™
3626914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
MDL Number MFCD00151450
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Odor Odorless
Quantity 5g
Assay Percent Notes (metals basis excluding Co), Co 0.1 to 1%
Health Hazard 2 GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Particle Size 10mm & down
Assay 99.5%
UN Number UN3077
Chemical Name or Material Nickel arsenide
CAS 12068-61-0
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.