Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel antimonide, 99.5% (metals basis excluding Co), Co 0.1-1%

Chemical Identifiers

Chemical Name or Material Nickel antimonide
CAS 12035-52-8
Formula Weight 180.46
MDL Number MFCD00050100
UN Number UN1549
Beilstein 84272
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3626709

  SDS
alfa aesar™
3626709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3626718

  SDS
alfa aesar™
3626718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 180.46
Health Hazard 1 Danger
Beilstein 84272
Health Hazard 3 GHS P Statement
P284a-P261-P280-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1549
Particle Size 10mm & down
MDL Number MFCD00050100
Chemical Name or Material Nickel antimonide
Assay 99.5%
Assay Percent Notes (metals basis excluding Co), Co 0.1 to 1%
Health Hazard 2 GHS H Statement
H331-H334-H351-H302-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
TSCA TSCA
Quantity 10g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
CAS 12035-52-8
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H351-H302-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.