missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel ammonium sulfate hexahydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01251922

 View more versions of this product

Catalog No. AA1251922



Description

Description

It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel ammonium sulfate hexahydrate
7785-20-8
100 g
MFCD00150260
Soluble in ammonium sulphate. Insoluble in alcohol. Slightly soluble in water.
Odorless
1.923
Green
Large Crystals
Ni(NH4)2(SO4)2·6H2O
Ammonium nickel sulfate hexahydrate
394.99 (286.90 Anhydrous)
Green crystals

Chemical Identifiers

7785-20-8
Ni(NH4)2(SO4)2·6H2O
MFCD00150260
Ammonium nickel sulfate hexahydrate
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H341-H350i-H360D-H372

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 239-793-5

RTECSNumber : WS6061000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only