Learn More
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1844136
Description
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Nickel ammonium sulfate hexahydrate | |
500 g | |
Ni(NH4)2(SO4)2·6H2O | |
UN3077 | |
Ammonium nickel sulfate hexahydrate | |
394.99 (286.90 Anhydrous) | |
Odorless |
7785-20-8 | |
H8N2NiO8S2·6H2O | |
MFCD00150260 | |
14,6501 | |
Soluble in water. | |
98% | |
1.923 |
Chemical Identifiers
7785-20-8 | |
H8N2NiO8S2·6H2O | |
MFCD00150260 | |
Ammonium nickel sulfate hexahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H334-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 239-793-5
RTECSNumber : WS6061000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only