Alfa Aesar™ Nickel Aluminum, Raney™ type non-activated
Manufacturer:  Alfa Aesar™ 8767636
 View more versions of this product
Catalog No. AA8767636
Specifications
Specifications
Ni:Al
500g
12635-29-9
MFCD00167392
aluminium-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminium nickel
Odorless
aluminum;nickel
6336846
Black to Gray
Nickel Aluminum
Raney™ type non-activated
AlNi
UN3208
NPXOKRUENSOPAO-UHFFFAOYSA-N
[Al].[Ni]
85.675
Powder
Chemical Identifiers
12635-29-9
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
6336846
[Al].[Ni]
AlNi
MFCD00167392
aluminium-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminium nickel
aluminum;nickel
Safety and Handling
Safety and Handling
GHS H Statement
H228-H261-H372-H351-H317
Flammable solid.
In contact with water releases flammable gas.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P210-P231+P232-P260-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
SDS Documents