missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel aluminum, 99.0% min (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 2289009

 View more versions of this product

Catalog No. AA2289009



Specifications

Specifications

Nickel aluminum
10g
12003-78-0
99.0% min. (metals basis)
MFCD00167392
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum;nickel
6336846
Gray
Gray powder, Intermetallic
Odorless
AlNi
aluminium-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminium nickel
[Al].[Ni]
85.675
Intermetallic Powder

Chemical Identifiers

12003-78-0
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
6336846
[Al].[Ni]
AlNi
MFCD00167392
aluminium-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminium nickel
aluminum;nickel
Safety and Handling

Safety and Handling

H334-H317-H351: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of causing cancer.

EINECSNumber : 234-439-6

SDS Documents