Learn More
Nickel aluminum, 99.0% min (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02289030
Description
Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
12003-78-0 | |
99.0% min. (metals basis) | |
MFCD00167392 | |
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel | |
NPXOKRUENSOPAO-UHFFFAOYSA-N | |
aluminum;nickel | |
6336846 | |
Powder |
Nickel aluminum | |
Gray powder, Intermetallic | |
AlNi | |
250 g | |
Insoluble in water. | |
[Al].[Ni] | |
85.675 | |
Odorless |
Chemical Identifiers
12003-78-0 | |
85.675 | |
NPXOKRUENSOPAO-UHFFFAOYSA-N | |
6336846 | |
[Al].[Ni] |
AlNi | |
MFCD00167392 | |
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel | |
aluminum;nickel |
Safety and Handling
P201-P202-P223-P231+P232-P281-P308+P313-P335+P334-P370+P378q-P501c
H261-H350
DOTInformation : Hazard Class: 4.3; Packaging Group: II
EINECSNumber : 234-439-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only