Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Aluminum

Wear resistant coatings

$83.01 - $83.01

Chemical Identifiers

Chemical Name or Material Nickel Aluminum
CAS 12635-27-7
MDL Number MFCD00167392
UN Number UN3208
Synonym Aluminum Nickel
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982822

  SDS
alfa aesar™
3982822
100g Each for $83.01

AA3982836

  SDS
alfa aesar™
3982836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
TSCA TSCA
MDL Number MFCD00167392
CAS 12635-27-7
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
Physical Form Powder
UN Number UN3208
Synonym Aluminum Nickel
Health Hazard 1 Danger
Quantity 100g
Chemical Name or Material Nickel Aluminum
Health Hazard 2 GHS H Statement
H228-H261-H372-H351-H317
Flammable solid.
In contact with water releases flammable gas.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Color Gray to black
Avg. Mol. Wt. or Mol. Wt. Range Ni:Al; 50:50 wt%
Health Hazard 3 GHS P Statement
P210-P231+P232-P260-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P260-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H261-H372-H351-H317
Flammable solid.
In contact with water releases flammable gas.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.