Please login to your online account to display your discounted pricing

Nickel AA Standard, 1mL = 1mg Ni (1,000ppm Ni), Ricca Chemical

$66.88 - $185.24

Chemical Identifiers

Chemical Name or Material Nickel AA Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.76
CAS Max % 4.95
CAS 7440-02-0
CAS Min % 0.09
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ANI1KN-100

 
ricca chemical company
RIANI1KN100
Poly natural 100mL Each for $66.88

ANI1KN-500

 
ricca chemical company
RIANI1KN500
Poly natural 500mL Each for $185.24
Description & Specifications

Specifications

Physical Form Liquid
Color Clear light green
Quantity 100mL
Equivalence 1 mL = 1 mg Ni
Concentration or Composition Notes 1,000ppm Ni
Packaging Poly natural
Chemical Name or Material Nickel AA Standard
CAS Min % 0.09
CAS Max % 4.95
CAS Max % 0.1
CAS Max % 97.0
CAS Min % 93.0
CAS 7732-18-5
CAS Min % 4.76
CAS 7697-37-2
CAS 7440-02-0