Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material Nickel 2-methoxyethoxide
CAS 142600-62-2
Linear Formula Ni(OCH2CH2OCH3)2
Formula Weight 208.76
MDL Number MFCD01863472
UN Number UN1188
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42378AC

 
alfa aesar™
42378AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42378AE

 
alfa aesar™
42378AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.76
Chemical Name or Material Nickel 2-methoxyethoxide
Packaging ChemSeal™ bottles
Sensitivity Moisture Sensitive
Physical Form Liquid
Linear Formula Ni(OCH2CH2OCH3)2
Concentration 5% w/v in 2-methoxyethanol
MDL Number MFCD01863472
CAS 142600-62-2
UN Number UN1188
Quantity 25mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H351-H226-H302-H312-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H351-H226-H302-H312-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

  • Packaged under Argon