Please login to your online account to display your discounted pricing

Nickel, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$112.40 - $423.07

Chemical Identifiers

Chemical Name or Material Nickel, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Ni
Formula Weight 58.69
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003447

 
avantor performance
577004
Poly Bottle 100mL Each for $423.07

02003448

 
avantor performance
646104
Narrow Mouth Poly Bottle 150mL Each for $112.40
Description & Specifications

Specifications

Formula Weight 58.69
Chemical Name or Material Nickel, 1000μg/mL (0.10% w/v)
Molecular Formula Ni
Quantity 100mL
Name Note Plasma Standard
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Purity Grade BAKER INSTRA-ANALYZED™ Reagent