missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niceritrol, TCI America™

$249.03 - $747.09

Chemical Identifiers

CAS 5868-05-3
Molecular Formula C29H24N4O8
Molecular Weight (g/mol) 556.53
MDL Number MFCD00215982
InChI Key KUEUWHJGRZKESU-UHFFFAOYSA-N
Synonym Pentaerythritol Tetranicotinate
PubChem CID 4476
IUPAC Name [3-(pyridine-3-carbonyloxy)-2,2-bis(pyridine-3-carbonyloxymethyl)propyl] pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)OCC(COC(=O)C2=CN=CC=C2)(COC(=O)C3=CN=CC=C3)COC(=O)C4=CN=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N097425MG View Documents TCI America
N097425MG
25mg Each for $249.03
 
N0974100MG View Documents TCI America
N0974100MG
100mg Each for $747.09
 
Specifications

Chemical Identifiers

5868-05-3
556.53
KUEUWHJGRZKESU-UHFFFAOYSA-N
4476
C1=CC(=CN=C1)C(=O)OCC(COC(=O)C2=CN=CC=C2)(COC(=O)C3=CN=CC=C3)COC(=O)C4=CN=CC=C4
C29H24N4O8
MFCD00215982
Pentaerythritol Tetranicotinate
[3-(pyridine-3-carbonyloxy)-2,2-bis(pyridine-3-carbonyloxymethyl)propyl] pyridine-3-carboxylate

Specifications

White-Yellow
25mg
C29H24N4O8
Pentaerythritol Tetranicotinate
C1=CC(=CN=C1)C(=O)OCC(COC(=O)C2=CN=CC=C2)(COC(=O)C3=CN=CC=C3)COC(=O)C4=CN=CC=C4
556.53
Crystal-Powder at 20°C
Niceritrol
165°C
5868-05-3
MFCD00215982
KUEUWHJGRZKESU-UHFFFAOYSA-N
[3-(pyridine-3-carbonyloxy)-2,2-bis(pyridine-3-carbonyloxymethyl)propyl] pyridine-3-carboxylate
4476
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

RTECSNumber : QT1980000

TSCA : No