missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nicardipine Hydrochloride, TCI America™

$158.89 - $462.92

Chemical Identifiers

CAS 54527-84-3
Molecular Formula C26H30ClN3O6
Molecular Weight (g/mol) 515.991
MDL Number MFCD00057327
InChI Key AIKVCUNQWYTVTO-UHFFFAOYSA-N
Synonym nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
PubChem CID 41114
IUPAC Name 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06355G View Documents TCI America
N06355G
5g Each for $158.89
 
N063525G View Documents TCI America
N063525G
25g Each for $462.92
 
Specifications

Chemical Identifiers

54527-84-3
515.991
AIKVCUNQWYTVTO-UHFFFAOYSA-N
41114
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
C26H30ClN3O6
MFCD00057327
nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride

Specifications

Undesignated
54527-84-3
MFCD00057327
nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
515.991
515.99
≥98.0% (HPLC,T)
5g
C26H30ClN3O6
2811
AIKVCUNQWYTVTO-UHFFFAOYSA-N
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
41114
Crystal-Powder at 20°C
Nicardipine Hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : US7972100

TSCA : No