missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nicardipine hydrochloride, 98%

L-type calcium channel blocker

Manufacturer:  Alfa Aesar™ J6126906

 View more versions of this product

Catalog No. AAJ6126906



Specifications

Specifications

Nicardipine hydrochloride
5g
C26H30ClN3O6
UN2811
angioglebil, bionicard, cardene, loxen, nicardil, nicardipine hcl, nicardipine hydrochloride, nicodel, perdipina, perdipine
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
515.991
515.99
98%
Yellow
54527-84-3
MFCD00057327
14,6495
AIKVCUNQWYTVTO-UHFFFAOYSA-N
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
41114
Powder

Chemical Identifiers

54527-84-3
515.991
AIKVCUNQWYTVTO-UHFFFAOYSA-N
41114
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
C26H30ClN3O6
MFCD00057327
angioglebil, bionicard, cardene, loxen, nicardil, nicardipine hcl, nicardipine hydrochloride, nicodel, perdipina, perdipine
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c

H301+H311-H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 259-198-4

RTECSNumber : US7972100

TSCA : No

Recommended Storage : Keep cold

Documents