Please login to your online account to display your discounted pricing

Niacin, U.S.P. - F.C.C., J.T.Baker™

$139.20 - $2,200.75

Chemical Identifiers

Chemical Name or Material Niacin
CAS 59-67-6
Purity Grade FCC/USP
Molecular Formula C6H5NO2
Formula Weight 123.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003519

 
avantor performance
274503
Wide Mouth Amber Glass Bottle 25g Each for $139.20

02003520

 
avantor performance
274501
Wide Mouth PVC Coated Glass Bottle 500g Each for $2,200.75
Description & Specifications

Specifications

Purity Grade FCC/USP
Formula Weight 123.11
Chemical Name or Material Niacin
Molecular Formula C6H5NO2
CAS 59-67-6
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.47kg
Quantity 25g