missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[NH2Me2][(RuCl((R)-tolbinap))2(mu-Cl)3], TCI America™

$74.86 - $279.59

Chemical Identifiers

CAS 749935-02-2
Molecular Formula C98H94Cl5NP4Ru2+2
Molecular Weight (g/mol) 1789.122
MDL Number MFCD09753034
InChI Key KXTUTRXHNWQYSZ-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131674756
IUPAC Name trichloranium;[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10171G View Documents TCI America
N10171G
1g Each for $279.59
 
N1017200MG View Documents TCI America
N1017200MG
200mg Each for $74.86
 
Specifications

Chemical Identifiers

749935-02-2
1789.122
KXTUTRXHNWQYSZ-UHFFFAOYSA-M
131674756
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru
C98H94Cl5NP4Ru2+2
MFCD09753034
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
trichloranium;[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride

Specifications

Undesignated
749935-02-2
MFCD09753034
KXTUTRXHNWQYSZ-UHFFFAOYSA-M
trichloranium;[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
131674756
Crystal-Powder at 20°C
1g
C98H94Cl5NP4Ru2+2
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]diruthenate(II)
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.CNC.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru
1789.122
1783.07
[NH2Me2][(RuCl((R)-tolbinap))2(mu-Cl)3]
Safety and Handling

Safety and Handling

TSCA : No