missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[NH2Me2][(RuCl((R)-segphos(regR)))2(mu-Cl)3], TCI America™

$93.20 - $293.34

Chemical Identifiers

CAS 346457-41-8
Molecular Formula C78H70Cl5NO8P4Ru2+2
Molecular Weight (g/mol) 1652.702
MDL Number MFCD09753037
InChI Key NDMWNTKVBQPQNJ-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
PubChem CID 131674758
IUPAC Name trichloranium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
SMILES CNC.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10211G View Documents TCI America
N10211G
1g Each for $293.34
 
N1021200MG View Documents TCI America
N1021200MG
200mg Each for $93.20
 
Specifications

Chemical Identifiers

346457-41-8
1652.702
NDMWNTKVBQPQNJ-UHFFFAOYSA-M
131674758
CNC.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru]
C78H70Cl5NO8P4Ru2+2
MFCD09753037
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
trichloranium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride

Specifications

Undesignated
346457-41-8
MFCD09753037
NDMWNTKVBQPQNJ-UHFFFAOYSA-M
trichloranium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
131674758
Crystal-Powder at 20°C
1g
C78H70Cl5NO8P4Ru2+2
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
CNC.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru]
1652.702
1646.65
[NH2Me2][(RuCl((R)-segphos(regR)))2(mu-Cl)3]
Safety and Handling

Safety and Handling

TSCA : No