missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[NH2Me2][(RuCl((R)-dm-segphos(regR)))2(mu-Cl)3], TCI America™
$97.89 - $336.78
Chemical Identifiers
CAS | 935449-46-0 |
---|---|
Molecular Formula | C94H96Cl5NO8P4Ru2 |
Molecular Weight (g/mol) | 1871.086 |
MDL Number | MFCD09753038 |
InChI Key | ICOHGSVVYSKYTD-UHFFFAOYSA-J |
Synonym | Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole]diruthenate(II) |
PubChem CID | 131675155 |
IUPAC Name | [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride |
SMILES | CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl- |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
N1023200MG
|
TCI America
N1023200MG |
200 mg |
Each for $97.89
|
|
|||||
N10231G
|
TCI America
N10231G |
1 g |
Each for $336.78
|
|
|||||
Chemical Identifiers
935449-46-0 | |
1871.086 | |
ICOHGSVVYSKYTD-UHFFFAOYSA-J | |
131675155 | |
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl- |
C94H96Cl5NO8P4Ru2 | |
MFCD09753038 | |
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole]diruthenate(II) | |
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride |
Specifications
935449-46-0 | |
MFCD09753038 | |
Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole]diruthenate(II) | |
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[Cl-].[Cl- | |
1871.086 | |
1871.09 | |
[NH2Me2][(RuCl((R)-dm-segphos(regR)))2(mu-Cl)3] |
C94H96Cl5NO8P4Ru2 | |
200 mg | |
ICOHGSVVYSKYTD-UHFFFAOYSA-J | |
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride | |
131675155 | |
Crystalline Powder |
Safety and Handling
TSCA : No