missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NGFR Mouse, PE, Clone: ME20.4, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $404.55

Specifications

Antigen NGFR
Clone ME20.4
Host Species Murine
Species Reactivity Canine, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Sheep
Applications Flow Cytometry,Microscopy,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122437 Life Technologies
LS12940041
25 Tests Each for $187.05
501122438 Life Technologies
LS12940042
100 Tests Each for $404.55
Specifications

Description

The monoclonal antibody ME20.4 recognizes CD271, a 75 kDa, type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesenchymal stem/stromal cells, neural crest stem cells, astrocytes, oligodendrocytes, and a subset of lymphoid cells including follicular dendritic cells. CD271 binds to the ligands NGF, BDNF, NTF3, and NTF4. CD271 contains a death domain which induces apoptosis and regulates cell growth and proliferation through interactions with TRAF2, TRAF4, TRAF6, PTPN13, and RANBP9.

The ME20.4 monoclonal antibody recognizes human, non-human primate, mouse, rat, sheep, rabbit, cat, dog, and pig CD271.
Description & Specifications

Specifications

NGFR
Murine
Flow Cytometry,Microscopy,Western Blot
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
ME20.4
Canine, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Sheep
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer