missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NGD 98-2 hydrochloride, Tocris™

High affinity CRF1 antagonist

$251.85 - $1,045.45

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C19H24F3N3O3·HCl
Formula Weight 435.87
Synonym N-(1-Ethylpropyl)-3-methoxy-5-[2-methoxy-4-(trifluoromethoxy)phenyl]-6-methyl-2-pyrazinamine hydrochloride
Chemical Name or Material NGD 98-2 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
431550 R & D Systems
431550
50mg Each for $1,045.45
431510 R & D Systems
431510
10mg Each for $251.85
Specifications Description & Specifications

Chemical Identifiers

>98%
435.87
NGD 98-2 hydrochloride
C19H24F3N3O3·HCl
N-(1-Ethylpropyl)-3-methoxy-5-[2-methoxy-4-(trifluoromethoxy)phenyl]-6-methyl-2-pyrazinamine hydrochloride

Specifications

50mg
C19H24F3N3O3·HCl
N-(1-Ethylpropyl)-3-methoxy-5-[2-methoxy-4-(trifluoromethoxy)phenyl]-6-methyl-2-pyrazinamine hydrochloride
>98%
435.87
NGD 98-2 hydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature