Please login to your online account to display your discounted pricing

NGD 94-1, Tocris

Selective D4 receptor ligand

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material NGD 94-1
Assay Percent Range >99%
Molecular Formula C18H20N6
Formula Weight 320.39
Synonym 2-[4-[(2-Phenyl-1H-imidazol-5-yl)methyl]-1-piperazinyl]-pyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

329810

 
r & d systems
329810
10mg Each for $201.36

329850

 
r & d systems
329850
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 320.39
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C18H20N6
Chemical Name or Material NGD 94-1
CAS 178928-68-2
Synonym 2-[4-[(2-Phenyl-1H-imidazol-5-yl)methyl]-1-piperazinyl]-pyrimidine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C