Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ NFκ B/p65 (Rel A), Rabbit Polyclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ NFκB/p65 (Rel A), Rabbit Polyclonal Antibody.

$277.63 - $1,407.29

Specifications

Antigen NFkappaB/p65 (Rel A)
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Format Quantity Price Quantity    

RB9034P

 
thermo scientific™ lab vision™
RB9034P
Concentrated 1mL Each for $1,407.29

RB9034P0

 
thermo scientific™ lab vision™
RB9034P0
Concentrated 100µL Each for $277.63

RB9034P1

 
thermo scientific™ lab vision™
RB9034P1
Concentrated 500µL Each for $841.71

RB9034R7

 
thermo scientific™ lab vision™
RB9034R7
Prediluted 7mL Each for $770.06
Description & Specifications

Specifications

Antigen NFkappaB/p65 (Rel A)
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Human
Cross Reactivity Rabbit
Host Species Rabbit
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Immunogen A synthetic peptide corresponding to internal domain of human NFkappaB p65
Isotype IgG

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ NFκB/p65 (Rel A), Rabbit Polyclonal Antibody.

NFκB is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IκB. In unstimulated cells, nuclear localization signals present on p65 are masked by members of the IκB family of inhibitory proteins. Subsequent to cell stimulation, IκB undergoes phosphorylation, ubiquitination and degradation by a proteosome-dependent pathway, allowing nuclear translocation of the active dimeric NFκB transcription factor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat and Rabbit. Others not tested.

Epitope: Internal domain

Immunogen: A synthetic peptide corresponding to internal domain of human NFκB/p65

Molecular Weight: 65kDa

Positive Control: Prostate gland and placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO