Please login to your online account to display your discounted pricing

NF 546, Tocris™

Selective P2Y11 agonist

$251.85 - $251.85

Chemical Identifiers

Chemical Name or Material NF 546
Assay Percent Range >95%
Molecular Formula C47H44N6Na4O17P4
Formula Weight 1180.74
Synonym 4,4'-(Carbonylbis(imino-3,1-phenylene-carbonylimino-3,1-(4-methyl-phenylene)carbonylimino))-bis(1,3-xylene-alpha,alpha'-diphosphonic acid tetrasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

389210

 
r & d systems
RDS389210
10mg Each for $251.85
Description & Specifications

Specifications

Formula Weight 1180.74
Quantity 10mg
Chemical Name or Material NF 546
Assay Percent Range >95%
Molecular Formula C47H44N6Na4O17P4
CAS 1006028-37-0
Synonym 4,4'-(Carbonylbis(imino-3,1-phenylene-carbonylimino-3,1-(4-methyl-phenylene)carbonylimino))-bis(1,3-xylene-alpha,alpha'-diphosphonic acid tetrasodium salt
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C