Please login to your online account to display your discounted pricing

NF 449, Tocris

Highly selective P2X1 antagonist

$264.88 - $1,099.17

Chemical Identifiers

Chemical Name or Material NF 449
Assay Percent Range >98%
Molecular Formula C41H24N6Na8O29S8
Formula Weight 1505.06
Synonym 4,4',4'',4'''-[Carbonylbis(imino-5,1,3-benzenetriyl-bis(carbonylimino))]tetrakis-1,3-benzenedisulfonic acid, octasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

139110

 
r & d systems
RDS139110
10mg Each for $264.88

139150

 
r & d systems
RDS139150
50mg Each for $1,099.17
Description & Specifications

Specifications

Formula Weight 1505.06
Quantity 10mg
Chemical Name or Material NF 449
CAS 627034-85-9
Synonym 4,4',4'',4'''-[Carbonylbis(imino-5,1,3-benzenetriyl-bis(carbonylimino))]tetrakis-1,3-benzenedisulfonic acid, octasodium salt
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C41H24N6Na8O29S8
Safety and Handling

Store at Room Temperature