Please login to your online account to display your discounted pricing

NF 279, Tocris


Potent and selective P2X1 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS119910

 View more versions of this product

Catalog No. 119910

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material NF 279
Quantity 10mg
CAS 202983-32-2
Recommended Storage Desiccate at -20°C
Assay Percent Range >98%
Molecular Formula C49H30N6Na6O23S6
Formula Weight 1401.1
Synonym 8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt

Specifications

Chemical Name or Material NF 279
Assay Percent Range >98%
Molecular Formula C49H30N6Na6O23S6
Formula Weight 1401.1
Synonym 8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
Accessories

Desiccate at -20°C