missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NF 279, Tocris

Potent and selective P2X1 antagonist

$232.08 - $956.46

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C49H30N6Na6O23S6
Formula Weight 1401.1
Synonym 8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
Chemical Name or Material NF 279
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
119950 R & D Systems
RDS119950
50mg Each for $956.46
119910 R & D Systems
RDS119910
10mg Each for $232.08
Specifications Description & Specifications

Chemical Identifiers

>98%
1401.1
NF 279
C49H30N6Na6O23S6
8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt

Specifications

50mg
>98%
1401.1
NF 279
202983-32-2
C49H30N6Na6O23S6
8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt

Safety and Handling

Recommended Storage : Desiccate at -20°C