Please login to your online account to display your discounted pricing

NF 157, Tocris

Selective P2Y11/P2X1 antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material NF 157
Assay Percent Range >98%
Molecular Formula C49H28F2N6Na6O23S6
Formula Weight 1437.08
Synonym 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-fluoro-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

245010

 
r & d systems
RDS245010
10mg Each for $257.85

245050

 
r & d systems
RDS245050
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 1437.08
Quantity 10mg
Chemical Name or Material NF 157
Assay Percent Range >98%
Molecular Formula C49H28F2N6Na6O23S6
CAS 104869-26-3
Synonym 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-fluoro-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature