Please login to your online account to display your discounted pricing

NF 110, Tocris

Potent P2X3 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material NF 110
Assay Percent Range >98%
Molecular Formula C41H28N6Na4O17S4
Formula Weight 1096.9
Synonym 4,4',4'',4'''-[Carbonylbis[imino-5,1,3-benzenetriylbis(carbonylimino)]]tetrakisbenzenesulfonic acid tetrasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

254850

 
r & d systems
RDS254850
50mg Each for $976.15

254810

 
r & d systems
RDS254810
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 1096.9
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C41H28N6Na4O17S4
CAS 111150-22-2
Chemical Name or Material NF 110
Synonym 4,4',4'',4'''-[Carbonylbis[imino-5,1,3-benzenetriylbis(carbonylimino)]]tetrakisbenzenesulfonic acid tetrasodium salt
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature