Please login to your online account to display your discounted pricing

NF 023, Tocris


Selective, competitive P2X1 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS124010

 View more versions of this product

Catalog No. 124010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 1162.86
Quantity 10mg
Chemical Name or Material NF 023
Assay Percent Range >98%
Molecular Formula C35H20N4Na6O21S6
CAS 104869-31-0
Synonym 8,8'-[carbonylbis(imino-3,1-phenylenecarbonylimino)]bis-1,3,5-naphthalene-trisulphonic acid, hexasodium salt
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material NF 023
Assay Percent Range >98%
Molecular Formula C35H20N4Na6O21S6
Formula Weight 1162.86
Synonym 8,8'-[carbonylbis(imino-3,1-phenylenecarbonylimino)]bis-1,3,5-naphthalene-trisulphonic acid, hexasodium salt
Accessories

Desiccate at Room Temperature