Please login to your online account to display your discounted pricing
Newtex™ Zetex™ and ZetexPlus™ High Temperature Gloves
Provide protection in temperatures to 1000°F
$94.04 - $1,407.96
Specifications
Coating Material None
Material Kevlar™ Fiberglass Fabric
For Use With (Application) foundries and casting operations, welding and hot works operations, Glass and Ceramic plants production, power generation, Petrochemical and refining, laboratory and testing facilities
Liner Material Wool
Temperature (English) Max. 1000°F
View More Specs
Products
Catalog Number Mfr. No. Temperature (English) Max. Length (English) Length (Metric) Price Quantity    
11-392-15
newtex industries inc
2100005
1000°F 11 in. 28cm Case of 12 Pair for $927.29
11-392-50
newtex industries inc
2100011
2000°F 11 in. 28cm Pair for $110.91
Case of 12 Pair for $1,215.24
11-392-52
newtex industries inc
2100012
2000°F 14 in. 35.5cm Case of 12 Pair for $1,407.96
11-392-51
newtex industries inc
2100006
1000°F 14 in. 35.5cm Pair for $94.04
Case of 12 Pair for $1,030.44
11-392-53
newtex industries inc
2100014
2000°F 23 in. 58.5cm Pair for $154.72
Case of 8 Pair for $1,237.76
11-392-59
newtex industries inc
2100007
1000°F 23 in. 58.5cm Pair for $195.96
Case of 8 Pair for $1,567.68
Description & Specifications
Specifications
Coating Material None
Material Kevlar™ Fiberglass Fabric
For Use With (Application) foundries and casting operations, welding and hot works operations, Glass and Ceramic plants production, power generation, Petrochemical and refining, laboratory and testing facilities
Liner Material Wool
Temperature (English) Max. 1000°F
Temperature (Metric) Max 537.7°C
  • Made from 100% premium Zetex texturized fiberglass fabric
  • Double stitched and reinforced
  • Premium wool liner