Please login to your online account to display your discounted pricing

NewPath Learning™ Interactive Whiteboard Science Lessons: Life Science


Manufacturer: fisher science education  54-6207

 View more versions of this product

Catalog No. S02300

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Specifications

Certifications/Compliance Smart™, Promethean™
Grade 6 to 8
License Single user
Classification Science
For Use With (Equipment) Computer/interactive whiteboard within a single school building
Compatibility Windows™, Mac™
For Use With (Application) Multisensory learning, they're the perfect resource for Interactive whiteboards, perfect complement to any biology curriculum- use on the whiteboard for the entire class or on a computer for an individual student
Product Type Chromosomes, Genes and DNA Multimedia Lesson
Includes 5 Modules, digital inquiry based student activity guide