Please login to your online account to display your discounted pricing

NewPath Learning™ Curriculum Mastery™ Games


Comprehensive coverage of the current National Science Education Standards.

Manufacturer: fisher science education  24-4001

 View more versions of this product

Catalog No. S95644

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Category Science
Product Type Curriculum Mastery™ Game
Grade 4
For Use With (Application) Comprehensive coverage of the current national science education standards
Includes 750 Review cards, teachers guide, classroom presentation CD