missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Curriculum Mastery™ Games

Comprehensive coverage of the current National Science Education Standards.

$231.25 - $231.25

Specifications

Category Science
For Use With (Application) Comprehensive coverage of the current national science education standards
Product Type Curriculum Mastery™ Game
Includes 750 Review cards, teachers guide, classroom presentation CD
View More Specs

Products 6
Catalog Number Mfr. No. Min. Grade Level Max. Grade Level Price Quantity  
Catalog Number Mfr. No. Min. Grade Level Max. Grade Level Price Quantity  
S95641 View Documents Fisher Scientific
241001
1 1 Each for $231.25
 
S95642 View Documents Fisher Scientific
242001
2 2 Each for $231.25
 
S95643 View Documents Fisher Scientific
243001
3 3 Each for $231.25
 
S95644 View Documents Fisher Scientific
244001
4 4 Each for $231.25
 
S95646 View Documents Fisher Scientific
246001
6 6 Each for $231.25
 
S95645 View Documents Fisher Scientific
245001
5 5 Each for $231.25
Case of 6 Each for $1,387.50
 
Description

Description

  • Class size: 36 students playing simultaneously
  • 25 sets of 30 standards-based review cards; 750 cards total
  • 12 double-sided, laminated game boards
  • Teacher planning and resource guide
  • Classroom presentation CD with interactive review questions for use on PC, Mac™ or Interactive Whiteboard
  • Introductory online subscription to access interactive activities and assessments

Specifications

Specifications

Science
Comprehensive coverage of the current national science education standards
Curriculum Mastery™ Game
750 Review cards, teachers guide, classroom presentation CD