missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NewPath Learning™ Curriculum Mastery™ Game: Mathematics

Class-pack or take-home editions available for Grades 1 to 8.

$113.40 - $113.40

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.



Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S95630 Fisher Science Education
23-2001
Each for $113.40
Case of 6 Each for $834.30
S95631 Fisher Science Education
23-3001
Each for $113.40
Case of 6 Each for $834.30
S95632 Fisher Science Education
23-4001
Each for $113.40
Case of 6 Each for $834.30
S95633 Fisher Science Education
23-5001
Each for $113.40
Case of 6 Each for $834.30
S95634 Fisher Science Education
23-6001
Each for $113.40
Case of 6 Each for $680.40
Specifications

Description

Comprehensive coverage of current NCTM focal points.

  • Includes 25 sets of 30 standards-based review cards and 12 double-sided laminated game boards — enough materials for 36 students playing simultaneously
  • Also includes Classroom Presentation CD with interactive review questions for use on Interactive Whiteboard or computer, and teacher guide.

Grade 1: Sequencing, Skip Counting, Number Line, Odd & Even, Shapes and more!
Grade 2: Place Value, Number Words, Fractions, Addition & Subtraction Facts and more!
Grade 3: Double Digit Addition & Subtraction, Estimation, Story Problems and more!
Grade 4: Rounding Numbers, Data Analysis, Graphs, Problem Solving and more!
Grade 5: Order of Operations, Common Factors, Fractions & Decimals, and more!
Grade 6: Distributive Property, Rational Numbers, Exponents, Areas, Formulas and more!
Grade 7: Graphing & Displaying Data, Plane Figures, Finding Volume, Algebra and more!
Grade 8: Equations & Inequalities, Polynomials, Geometry, Linear Equations and more!
Fractions & Decimals (Grades 3 to 6): Fractions, Decimals, Proportions, Percents and more!
Algebra (Grades 6 to 10): Commutative & Associative Properties, Equations, Functions and more!