Please login to your online account to display your discounted pricing

NewPath™ Curriculum Mastery™ Game: Science Skills Middle School

Comprehensive coverage of the current NSES standards.

$139.05 - $139.05

Specifications

Category Science
For Use With (Application) Comprehensive coverage of the current national science education standards
Grade 5 to High School
View More Specs
Products
Catalog Number Mfr. No. Includes Format Product Type Price Quantity    

S95685

 
fisher science education
24-6008
750 Review cards, teachers guide, classroom presentation CD Class-pack edition Earth Science game Each for $139.05
Case of 6 Each for $834.30

S95684

 
fisher science education
24-6007
750 Review cards, teachers guide, classroom presentation CD Class-pack edition Life Science game Each for $139.05
Case of 6 Each for $834.30

S95686

 
fisher science education
24-6009
750 Review cards, teachers guide, classroom presentation CD Class-pack edition Physical Science game Each for $139.05
Case of 6 Each for $834.30
Description & Specifications

Specifications

Category Science
For Use With (Application) Comprehensive coverage of the current national science education standards
Grade 5 to High School

Class-pack or take-home editions available.

Class-Pack Edition
Class size: 36 students playing simultaneously.

  • 25 Sets of 30 standards-based review cards, 750 cards total
  • 12 Double-sided, laminated game boards
  • Teacher planning and resource guide
  • Classroom presentation CD with interactive review questions for use on PC, Mac™, or projection unit
  • Introductory online subscription to access interactive activities and assessments
Take-Home Edition
Ideal for promoting parental involvement.
  • 25 Games, individually packaged in vinyl folders with a set of 30 standards-based review questions, a double-sided game board, and materials for four students
  • Teacher and parent guide
  • Classroom presentation CD
  • Online subscription

Life Science Skills: Cells: The Basic Units of Life; Cell Processes; Cell Reproduction; Genetics – Study of Heredity; Modern Genetics; The Evolution and Interaction of Living Things; Bacteria and Viruses; Protists and Fungi; Introduction to Plants; Introduction to Animals

Earth Science Skills: Introduction to Earth Science, Maps as Models of the Earth/Contour Models, Minerals, Rocks, Fossils, Earth's Energy Resources, Geologic Time, Plate Tectonics, Earthquakes, Volcanoes, Weathering of Rocks & Soil Formation, Eartht's Fresh Water,Groundwater Resources,Erosion & Deposition,Oceans/Oceanography, The Movement of Ocean Water,
Earth's Atmosphere, Understanding Weather, Weather Patterns, Climate, Studying and Exploring Space, Stars, Galaxies and the Universe, The Sun-Earth-Moon System, Our Solar System,Our Impact on Earth


Physical Science Skills:Introduction to Physical Science, Introduction to Matter,Solids, Liquids & Gases, Mixtures, Solutions & Compounds, Motion, Forces, Forces in Fluids, Work and Machines, Energy and Energy Resources, Heat and Heat Technology,Properties of Atoms, Elements and the Periodic Table, Chemical Bonding, Chemical Reactions, Acids, Bases and Salts, Organic Compounds, Electricity, Magnetism, Electromagnetism, The Energy of Waves, Sound, Light, Mirrors and Lenses, Chemistry in Our World, Technology in Our World